Läs mer
5-(Difluoromethyl)-2-pyridinamine (>80%), TRC
Produktkod. 30364959
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364959 10 mg
30350882 50 mg
30355556 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30364959 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD26163110MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1211523-68-0
Molekylformel C6H6N2F2
Molekylvikt (g/mol) 144.12
LEDER Nc1ccc(cn1)C(F)F
Specifikationer
Kemiskt namn eller material 5-(Difluoromethyl)-2-pyridinamine
Kvantitet 10 mg
InChI formel InChI=1S/C6H6F2N2/c7-6(8)4-1-2-5(9)10-3-4/h1-3,6H,(H2,9,10)