missing translation for 'onlineSavingsMsg'
Learn More

5-(Dihydroxyboryl)-2-thiophenecarboxylic acid, 95+%, Maybridge™

Catalog Number p-4510094
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
465515-31-5
C5H5BO4S
171.961
OQGIKNPOYTVNNF-UHFFFAOYSA-N
2-carboxythiophene-5-boronic acid, 5-carboxythiophene-2-boronic acid, 5-dihydroxyboryl-2-thiophenecarboxylic acid, 5-dihydroxyboryl thiophene-2-carboxylic acid, 5-dihydroxyboranyl thiophene-2-carboxylic acid, 5-borono-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-borono, pubchem7851, acmc-1ap1p, d0jv5j
2779807
5-boronothiophene-2-carboxylic acid
B(C1=CC=C(S1)C(=O)O)(O)O
This item is not returnable. View return policy
465515-31-5
C5H5BO4S
171.961
OQGIKNPOYTVNNF-UHFFFAOYSA-N
2-carboxythiophene-5-boronic acid, 5-carboxythiophene-2-boronic acid, 5-dihydroxyboryl-2-thiophenecarboxylic acid, 5-dihydroxyboryl thiophene-2-carboxylic acid, 5-dihydroxyboranyl thiophene-2-carboxylic acid, 5-borono-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-borono, pubchem7851, acmc-1ap1p, d0jv5j
2779807
5-boronothiophene-2-carboxylic acid
B(C1=CC=C(S1)C(=O)O)(O)O
Assay Percent Range 95%
Quantity 5 g
Formula Weight 171.97
Percent Purity 95%
Chemical Name or Material 5-(Dihydroxyboryl)-2-thiophenecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'