Läs mer
5-DOXYL-stearic Acid, Free Radical (>90%), TRC
Produktkod. 30011392
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011392 10 mg
30005718 5 mg
30004892 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011392 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD53552810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 29545-48-0
Molekylformel C22H42NO4
Molekylvikt (g/mol) 384.57
Synonym 2-(3-Carboxypropyl)-4,4-dimethyl-2-tridecyl-3-oxazolidinyloxy, 2-(3-Carboxypropyl)-2-tridecyl-4,4-dimethyl oxazoline-3-oxyl, 2-(3-Carboxypropyl)-4,4-dimethyl-2-tridecyl-3-oxazolidinyloxyl, 5-DSA, 5-Doxylstearate, 5-Doxylstearic acid, 5-Nitroxystearate, 5-SASL, 5NS, K 5D, K 5D (spin probe), 2-(3--Carboxypropyl)-4,4-dimethyl-2-tridecyl-3-oxazolidinyloxy
LEDER OS(C(F)(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)F)(=O)=O
Specifikationer
Kemiskt namn eller material 5-DOXYL-stearic Acid Free Radical
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
Procent renhet >95%