missing translation for 'onlineSavingsMsg'
Läs mer

5-Ethyl-1,3-benzenediol, TRC

Produktkod. 30138454
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138454 500 mg
30137079 1 g
30010478 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30138454 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE896700500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 4299-72-3
Molekylformel C8H10O2
Molekylvikt (g/mol) 138.16
Synonym 5-Ethyl-resorcinol, 5-Ethylresorcin, 5-Ethylresorcinol, Homoorcinol;
LEDER CCc1cc(O)cc(O)c1

Specifikationer

Kemiskt namn eller material 5-Ethyl-1,3-benzenediol
Kvantitet 500 mg
InChI formel InChI=1S/C8H10O2/c1-2-6-3-7(9)5-8(10)4-6/h3-5,9-10H,2H2,1H3
Formel vikt 138.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.