missing translation for 'onlineSavingsMsg'
Läs mer

5-Ethynyl-2’-deoxyuridine, TRC

Produktkod. 30369483
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30369483 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30369483 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE932175100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 5-Ethynyl-2'-deoxyuridine
CAS 61135-33-9
Molekylformel C11 H12 N2 O5
Kvantitet 100 mg
Synonym EdU, 2'-Deoxy-5-ethynyluridine
InChI formel InChI=1S/C11H12N2O5/c1-2-6-4-13(11(17)12-10(6)16)9-3-7(15)8(5-14)18-9/h1,4,7-9,14-15H,3,5H2,(H,12,16,17)/t7-,8+,9+/m0/s1
LEDER OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(C#C)C(=O)NC2=O
IUPAC-namn 5-ethynyl-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
Molekylvikt (g/mol) 252.22
Formel vikt 252.0746
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.