missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoro-2-hydroxy-3-nitropyridine, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Quantity | 250 mg |
Beilstein | 7582013 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 158.09 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 5-Fluoro-2-hydroxy-3-nitropyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'