missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylbenzoic acid, 99%, Thermo Scientific™

Catalog Number p-7080720
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
33184-16-6
C8H7FO2
154.14
MFCD00042294
JVBLXLBINTYFPR-UHFFFAOYSA-N
2-methyl-5-fluorobenzoic acid, 5-fluoro-o-toluic acid, 3-fluoro-6-methylbenzoic acid, 5-fluoro-2-methylbenzoicacid, benzoic acid, 5-fluoro-2-methyl, 5-fluoro-2-methyl-benzoic acid, 5-fluoro-2-methyl benzoic acid, benzoicacid, 5-fluoro-2-methyl, pubchem1326
182114
5-fluoro-2-methylbenzoic acid
CC1=C(C=C(C=C1)F)C(=O)O
This item is not returnable. View return policy
33184-16-6
C8H7FO2
154.14
MFCD00042294
JVBLXLBINTYFPR-UHFFFAOYSA-N
2-methyl-5-fluorobenzoic acid, 5-fluoro-o-toluic acid, 3-fluoro-6-methylbenzoic acid, 5-fluoro-2-methylbenzoicacid, benzoic acid, 5-fluoro-2-methyl, 5-fluoro-2-methyl-benzoic acid, 5-fluoro-2-methyl benzoic acid, benzoicacid, 5-fluoro-2-methyl, pubchem1326
182114
5-fluoro-2-methylbenzoic acid
CC1=C(C=C(C=C1)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 130°C to 132°C
Quantity 1 g
Beilstein 2249813
Formula Weight 154.14
Percent Purity 99%
Chemical Name or Material 5-Fluoro-2-methylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'