Läs mer
5-Fluoro-2-methylphenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific™
Produktkod. 15450936
Change view
Kvantitet:
100 mL
Förpackningsstorlek:
100ml
Produktkod. Kvantitet unitSize
15450936 100 mL 100ml
1 options
Produktkod. 15450936 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H54234.AE
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Fluoro-2-methylphenylmagnesium bromide
CAS 186496-59-3
Molekylformel C7H6BrFMg
MDL-nummer MFCD01311468
Kvantitet 100 mL
UN-nummer UN2924
Känslighet Air and moisture sensitive
Synonym bromo 5-fluoro-2-methylphenyl magnesium, magnesium,bromo 5-fluoro-2-methylphenyl, xkmsjpvajjfyfu-uhfffaoysa-m, 5-fluoro-2-methylphenylmagnesium bromide, 2-methyl-5-fluorophenyl magnesium bromide, 5-fluoro-2-methylphenylmagnesium bromide solution, 5-fluoro-2-methylphenylmagnesium bromide, 0.50 m in 2-methf, 5-fluoro-2-methylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 5-fluoro-2-methylphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
InChI-nyckel XKMSJPVAJJFYFU-UHFFFAOYSA-M
LEDER CC1=CC=C(F)C=C1[Mg]Br
IUPAC-namn magnesium;1-fluoro-4-methylbenzene-5-ide;bromide
Molekylvikt (g/mol) 213.33
PubChem CID 2778349
Formel vikt 213.33
Koncentration eller sammansättning (efter analyt eller komponenter) 0.50M in 2-MeTHF