Learn More
Alfa Aesar™ 5-Fluoro-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole, 97%
Brand:  Alfa Aesar™ H63579.MD
Additional Details : CAS Number : 116480-54-7 Weight : 0.00025kg
Quantity 250mg
 View more versions of this product
Product Code. 15448386
  • 648.87 SEK / 250mg
Estimated Shipment date
from Supplier 31-08-2018
Add to basket
Specifications
Chemical Identifiers
116480-54-7
230.286
XFERGYRFFJXSGI-UHFFFAOYSA-N
C14H15FN2
MFCD23380604
14973209
Specifications
5-Fluoro-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole
116480-54-7
C14H15FN2
11193308
CN1CCC(=CC1)C2=CNC3=C2C=C(C=C3)F
230.286
230.28
250mg
97%
MFCD23380604
XFERGYRFFJXSGI-UHFFFAOYSA-N
5-fluoro-3-(1-methyl-3,6-dihydro-2H-pyridin-4-yl)-1H-indole
14973209
Solid