Läs mer
5-Fluoro Cytosine-13C,15N2, TRC
Produktkod. 17954605
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17954605 10 mg
17983235 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17954605 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF58900210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1216616-31-7
Molekylformel 13C C3 H4 F 15N2 N O
Molekylvikt (g/mol) 132.07
Synonym 2(1H)-Pyrimidinone-2-13C-1,3-15N2, 6-amino-5-fluoro-, 4-Amino-5-fluoropyrimidin-2(1H)-one-2-13C-1,3-15N2, 6-Amino-5-fluoro-2(1H)-pyrimidinone-2-13C-1,3-15N2, Flucytosine-13C,15N2, 5-Fluorocytosine-2-13C-1,3-15N2, 5-Fluoro cytosine-13C,15N2, 6-Amino-2-oxo-5-fluoropyrimidine-2-13C-1,3-15N2
IUPAC-namn 4-amino-5-fluoro-(2^{13}C,1,3-^{15}N_{2})1H-pyrimidin-2-one
LEDER NC1=[15N][13C](=O)[15NH]C=C1F
Specifikationer
Kemiskt namn eller material 5-Fluorocytosine-2-13C-1,3-15N2
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C4H4FN3O/c5-2-1-7-4(9)8-3(2)6/h1H,(H3,6,7,8,9)/i4+1,7+1,8+1
Formel vikt 132.0313
Procent renhet >95%