Läs mer
5-Fluorodihydropyrimidine-2,4-dione-13C,15N2, TRC
Produktkod. 30003835
Change view
Kvantitet:
0.5 mg
5 mg
Förpackningsstorlek:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30003835 5 mg -
30011690 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30003835 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF5910275MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1189492-99-6
Molekylformel C313CH5F15N2O2
Molekylvikt (g/mol) 135.07
Synonym 5-Fluorodihydropyrimidine-2,4-Dione 13C,15N2
LEDER FC1C[15NH][13C](=O)[15NH]C1=O
Specifikationer
Kemiskt namn eller material 5-Fluorodihydropyrimidine-2,4-dione-13C,15N2
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C4H5FN2O2/c5-2-1-6-4(9)7-3(2)8/h2H,1H2,(H2,6,7,8,9)/i4+1,6+1,7+1
Formel vikt 135.0309
Procent renhet >95%