Läs mer
5-Fluorodihydropyrimidine-2,4-dione, TRC
Produktkod. 30009919
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009919 25 mg
30004642 10 mg
30007907 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009919 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF59102525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 696-06-0
Molekylformel C4 H5 F N2 O2
Molekylvikt (g/mol) 132.093
Synonym 5,6-Dihydro-5-fluorouracil, 2,4(1H,3H)-Pyrimidinedione, 5-fluorodihydro-, 5-Fluorodihydro-2,4(1H,3H)-pyrimidinedione (ACI), Hydrouracil, 5-fluoro- (6CI, 7CI, 8CI), 5,6-Dihydro-5-fluorouracil, 5-Fluoro-5,6-dihydrouracil, 5-Fluorodihydrouracil, 5-Fluorohydrouracil, Dihydro-5-fluorouracil
IUPAC-namn 5-fluoro-1,3-diazinane-2,4-dione
LEDER FC1CNC(=O)NC1=O
Specifikationer
Kemiskt namn eller material 5-Fluorodihydropyrimidine-2,4-dione
Kvantitet 25 mg
InChI formel InChI=1S/C4H5FN2O2/c5-2-1-6-4(9)7-3(2)8/h2H,1H2,(H2,6,7,8,9)
Formel vikt 132.034