missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindole-3-carboxaldehyde, 99%, Thermo Scientific™

Catalog Number p-4813218
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2338-71-8
C9H6FNO
163.15
MFCD00022719
YUAJKGBLPVLADK-UHFFFAOYSA-N
5-fluoroindole-3-carboxaldehyde, 5-fluoro-1h-indole-3-carboxaldehyde, 5-fluoroindole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 5-fluoro, 3-formyl-5-fluoroindole, pubchem7267, zlchem 1197, 5-fluoro-3-formylindole, 5-fluoroindole-3-aldehyde, acmc-209g2v
259089
5-fluoro-1H-indole-3-carbaldehyde
FC1=CC=C2NC=C(C=O)C2=C1
This item is not returnable. View return policy
2338-71-8
C9H6FNO
163.15
MFCD00022719
YUAJKGBLPVLADK-UHFFFAOYSA-N
5-fluoroindole-3-carboxaldehyde, 5-fluoro-1h-indole-3-carboxaldehyde, 5-fluoroindole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 5-fluoro, 3-formyl-5-fluoroindole, pubchem7267, zlchem 1197, 5-fluoro-3-formylindole, 5-fluoroindole-3-aldehyde, acmc-209g2v
259089
5-fluoro-1H-indole-3-carbaldehyde
FC1=CC=C2NC=C(C=O)C2=C1
Melting Point 170°C to 172°C
Assay Percent Range 98.5 % min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 163.15
Percent Purity 99%
Chemical Name or Material 5-Fluoroindole-3-carboxaldehyde
compliance-icons
Product Identifier
  • 5-Fluoroindole-3-carboxaldehyde, 99%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'