Läs mer
5-Fluorooxindole (5-Fluoro-1,3-dihydro-2H-indol-2-one), Mikromol
Produktkod. 30462046
Change view
Kvantitet:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30462046 4 x 25 mg
30050577 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30462046 Leverantör Mikromol Leverantörsnummer MM3568.0200254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Fluorooxindole (5-Fluoro-1,3-dihydro-2H-indol-2-one), Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Fluoro-2-oxindole
CAS 56341-41-4
Analyt- eller komponentnamn 5-Fluoro-2-oxindole
Frakt skick Room Temperature
Molekylformel C8 H6 F N O
Certifikat och efterlevnad ISO 17025
Synonym 5-Fluoro-1,3-dihydro-2H-indol-2-one, 5-Fluoro-1,3-dihydroindol-2-one, 5-Fluoro-2,3-dihydro-2-oxoindole, 5-Fluoro-2,3-dihydroindol-2-one, 5-Fluoro-2-oxindole, 5-Fluoro-2-oxoindoline, 5-Fluoroindolin-2-one, 5-Fluorooxindole
InChI formel InChI=1S/C8H6FNO/c9-6-1-2-7-5(3-6)4-8(11)10-7/h1-3H,4H2,(H,10,11)
Lösningstyp Neat
LEDER Fc1ccc2NC(=O)Cc2c1
IUPAC-namn 5-fluoro-1,3-dihydroindol-2-one
Molekylvikt (g/mol) 151.14
Kvantitet 4 x 25 mg
Formel vikt 151.0433 g/mol