Läs mer
5-Fluorouracil-13C,15N2, TRC
Produktkod. 30009516
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009516 10 mg
30012375 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009516 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF59600210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1189423-58-2
Molekylformel C313CH3F15N2O2
Molekylvikt (g/mol) 133.06
Synonym 5-Fluoro-2,4(1H,3H)pyrimidinedione-13C,15N2, 2,4-Dihydroxy-5 -fluoropyrimidine-13C,15N2, 5-FU-13C,15N2, 5-Fluoropyrimidine-2,4-diol-13C,15N2, Adrucil-13C,15N2, Arumel-13C,15N2, Carac-13C,15N2, Fluorouracil-13C,15N2, Queroplex-13C,15N2, Ro 2-9757-13C,15N2, Timazin-13C,15N2, U 8953-13C,15N2;
LEDER O=[13C]1[15NH]C=C(F)C([15NH]1)=O
Specifikationer
Kemiskt namn eller material 5-Fluorouracil-13C,15N2
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
Formel vikt 133.0153
Procent renhet >95%