Läs mer
5-Formylcytosine, TRC
Produktkod. 30137361
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137361 1 mg
30009050 10 mg
30003771 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137361 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF6989751MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4425-59-6
Molekylformel C5H5N3O2
Molekylvikt (g/mol) 139.11
Synonym 4-Amino-1,2-dihydro-2-oxo-5-pyrimidinecarboxaldehyde, 4-Amino-2-hydroxy-5-pyrimidinecarboxaldehyde, 6-Amino-1,2-dihydro-2-oxo-5-pyrimidinecarboxaldehyde, 4-Amino-5-formyl-2(1H)-pyrimidinone, 5-Formyl-6-amino-2,3-dihydro-2-oxopyrimidine
IUPAC-namn 6-amino-2-oxo-1,2-dihydropyrimidine-5-carbaldehyde
LEDER NC1=C(C=O)C=NC(N1)=O
Specifikationer
Kemiskt namn eller material 5-Formylcytosine
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C5H5N3O2/c6-4-3(2-9)1-7-5(10)8-4/h1-2H,(H3,6,7,8,10)
Formel vikt 139.04
Procent renhet >95%