Learn More
5-Hydroxy-2-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog Number p-7077150
Click to view available options
Quantity:
1 g
5 g
25 g
42454-06-8
C7H5NO4
167.12
MFCD00007332
XLYPHUGUKGMURE-UHFFFAOYSA-N
3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
39211
5-hydroxy-2-nitrobenzaldehyde
OC1=CC=C(C(C=O)=C1)[N+]([O-])=O
42454-06-8
C7H5NO4
167.12
MFCD00007332
XLYPHUGUKGMURE-UHFFFAOYSA-N
3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
39211
5-hydroxy-2-nitrobenzaldehyde
OC1=CC=C(C(C=O)=C1)[N+]([O-])=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 168.0°C to 170.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula HOC6H3(NO2)CHO
Quantity 1 g
Beilstein 08,63
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 5-Hydroxy-2-nitrobenzaldehyde
compliance-icons
Product Identifier
  • 5-Hydroxy-2-nitrobenzaldehyde, 98%
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.