Läs mer
5-Hydroxy-6-methyl-3,4-pyridinedicarboxylic Acid, TRC
Produktkod. 30138679
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138679 250 mg
17901235 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138679 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH946970250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 479-30-1
Molekylformel C8H7NO5
Molekylvikt (g/mol) 197.14
Synonym Ichiba Acid
IUPAC-namn 5-hydroxy-6-methylpyridine-3,4-dicarboxylic acid
LEDER O=C(C1=C(C(O)=O)C(O)=C(C)N=C1)O
Specifikationer
Kemiskt namn eller material 5-Hydroxy-6-methyl-3,4-pyridinedicarboxylic Acid
Kvantitet 250 mg
InChI formel InChI=1S/C8H7NO5/c1-3-6(10)5(8(13)14)4(2-9-3)7(11)12/h2,10H,1H3,(H,11,12)(H,13,14)
Formel vikt 197.0324