Läs mer
5-hydroxy Chlorosulfuron, TRC
Produktkod. 30180259
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180259 5 mg
30180234 1 mg
30179834 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180259 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH8394055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81123-38-8
Molekylformel C12 H12 Cl N5 O5 S
Molekylvikt (g/mol) 373.77
Synonym 2-Chloro-5-hydroxy-N-[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]benzenesulfonamide, N 5754
IUPAC-namn 1-(2-chloro-5-hydroxyphenyl)sulfonyl-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)urea
LEDER COc1nc(C)nc(NC(=O)NS(=O)(=O)c2cc(O)ccc2Cl)n1
Specifikationer
Kemiskt namn eller material Chlorsulfuron-5-hydroxy
Kvantitet 5 mg
InChI formel InChI=1S/C12H12ClN5O5S/c1-6-14-10(17-12(15-6)23-2)16-11(20)18-24(21,22)9-5-7(19)3-4-8(9)13/h3-5,19H,1-2H3,(H2,14,15,16,17,18,20)
Formel vikt 373.0248