Läs mer
5-Hydroxy Diclofenac, TRC
Produktkod. 30009567
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009567 1 mg
30012653 5 mg
30011403 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009567 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH8252301MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 69002-84-2
Molekylformel C14 H11 Cl2 N O3
Molekylvikt (g/mol) 312.15
Synonym Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-5-hydroxy-, 2-[(2,6-Dichlorophenyl)amino]-5-hydroxybenzeneacetic acid, 5-Hydroxydiclofenac, 5-Hydroxy Diclofenac
IUPAC-namn 2-[2-(2,6-dichloroanilino)-5-hydroxyphenyl]acetic acid
LEDER OC(=O)Cc1cc(O)ccc1Nc2c(Cl)cccc2Cl
Specifikationer
Kemiskt namn eller material 5-Hydroxy Diclofenac
Kvantitet 1 mg
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C14H11Cl2NO3/c15-10-2-1-3-11(16)14(10)17-12-5-4-9(18)6-8(12)7-13(19)20/h1-6,17-18H,7H2,(H,19,20)
Formel vikt 311.0116
Procent renhet >95%