Läs mer
5-Hydroxy Flunixin (Contain 5% Flunixin), TRC
Produktkod. 17971845
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17971845 2.5 mg 2.5mg
17911535 1 mg -
17924075 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17971845 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9424202.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75369-61-8
Molekylformel C14 H11 F3 N2 O3
Molekylvikt (g/mol) 312.24
Synonym 5-Hydroxy-2-[[2-methyl-3-(trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic acid, 5-Hydroxyflunixin
IUPAC-namn 5-hydroxy-2-[2-methyl-3-(trifluoromethyl)anilino]pyridine-3-carboxylic acid
LEDER Cc1c(Nc2ncc(O)cc2C(=O)O)cccc1C(F)(F)F
Specifikationer
Kemiskt namn eller material 5-Hydroxy Flunixin
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C14H11F3N2O3/c1-7-10(14(15,16)17)3-2-4-11(7)19-12-9(13(21)22)5-8(20)6-18-12/h2-6,20H,1H3,(H,18,19)(H,21,22)
Formel vikt 312.0722
Procent renhet >95%