Läs mer
5-Hydroxy L-Tryptophan, TRC
Produktkod. 30135133
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135133 100 mg
30004237 250 mg
30005123 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135133 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH975710100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4350-09-8
Molekylformel C11 H12 N2 O3
Molekylvikt (g/mol) 220.22
Synonym (S)-2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic Acid, N-Acetyltryptophan Imp. D (EP), Tryptophan Imp. D (EP), 5-Hydroxytryptophan, Tryptophan, 5-hydroxy-, L- (8CI), (S)-5-Hydroxytryptophan, 5-Hydroxy-L-tryptophan, 5-Hydroxyl-L-tryptophan, Cincofarm, L-5-HTP, L-5-Hydroxytryptophan, Levothym, Levotinine, Oxitriptan, Oxyfan, Pretonine, Quietim, Serotonyl, Telesol, Tript-Oh, Triptene
IUPAC-namn (2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
LEDER N[C@@H](Cc1c[nH]c2ccc(O)cc12)C(=O)O
Specifikationer
Kemiskt namn eller material 5-Hydroxy-L-tryptophan
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C11H12N2O3/c12-9(11(15)16)3-6-5-13-10-2-1-7(14)4-8(6)10/h1-2,4-5,9,13-14H,3,12H2,(H,15,16)/t9-/m0/s1
Kvantitet 100 mg
Formel vikt 220.0848
Procent renhet >95%