Läs mer
5-Hydroxy-N-methyl-2-pyrrolidinone, TRC
Produktkod. 30134643
Change view
Kvantitet:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134643 1 g
30006394 100 mg
30004710 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134643 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9483201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 41194-00-7
Molekylformel C5 H9 N O2
Molekylvikt (g/mol) 115.13
Synonym 2-Pyrrolidinone, 5-hydroxy-1-methyl-, 5-Hydroxy-1-methyl-2-pyrrolidinone, 5-Hydroxy-1-methylpyrrolidin-2-one, 5-Hydroxy-N-methyl-2-pyrrolidone, 5-Hydroxy-N-methylpyrrolidone
IUPAC-namn 5-hydroxy-1-methylpyrrolidin-2-one
LEDER CN1C(O)CCC1=O
Specifikationer
Kemiskt namn eller material 5-Hydroxy-N-methyl-2-pyrrolidinone
Kvantitet 1 g
InChI formel InChI=1S/C5H9NO2/c1-6-4(7)2-3-5(6)8/h4,7H,2-3H2,1H3
Formel vikt 115.0633