Läs mer
5-(Hydroxymethyl)-2,4-dimethylpyridin-3-ol Hydrochloride, Mikromol™
Beskrivning
5-(Hydroxymethyl)-2,4-dimethylpyridin-3-ol Hydrochloride
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-Deoxypyridoxine hydrochloride |
| CAS | 148-51-6 |
| Analyt- eller komponentnamn | 4-Deoxypyridoxine hydrochloride |
| Frakt skick | Ice pack (-20°C) |
| Molekylformel | C8 H11 N O2 . Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | 2,4-Dimethyl-3-hydroxy-5-hydroxymethylpyridine Hydrochloride, 4-Deoxypyridoxine Hydrochloride, 3-Hydroxy-5-hydroxymethyl-2,4-dimethylpyridine Hydrochloride, 4-Deoxypyridoxol Hydrochloride, 5-Hydroxy-4,6-dimethyl-3-pyridinemethanol Hydrochloride, 4-Desoxyadermin Hydrochloride, 4-Desoxypyridoxine Hydrochloride, 5-Hydroxymethyl-2,4-dimethylpyridin-3-ol Hydrochloride, Pyridoxine Hydrochloride Imp. B (EP), Pyridoxine Imp. B (EP), 5-(Hydroxymethyl)-2,4-dimethylpyridin-3-ol Hydrochloride, Pyridoxine Hydrochloride Imp. B (EP) as Hydrochloride, Pyridoxine Hydrochloride Impurity B as Hydrochloride, Pyridoxine Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C8H11NO2.ClH/c1-5-7(4-10)3-9-6(2)8(5)11;/h3,10-11H,4H2,1-2H3;1H |
| LEDER | Cl.Cc1ncc(CO)c(C)c1O |
| IUPAC-namn | 5-(hydroxymethyl)-2,4-dimethylpyridin-3-ol;hydrochloride |
| Visa mer |