Läs mer
5-Hydroxymethyl-2-furaldehyde-13C6, TRC
Produktkod. 30012139
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012139 10 mg
30006074 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012139 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94700210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1219193-98-2
Molekylformel 13C6H6O3
Molekylvikt (g/mol) 132.07
Synonym 5-(Hydroxymethyl)-2-furancarboxaldehyde-13C6, 2-Formyl-5-hydroxymethylfuran-13C6, HMF, 2-Hydroxymethyl-5-furfural-13C6, 5-(Hydroxymethyl)-2-furancarbonal-13C6, 5-(Hydroxymethyl)furfural-13C6, 5-Hydroxymethylfurfurol-13C6, 5-Oxymethylfurfurole-13C6, NSC 40738-13C6;
LEDER O[13CH2][13C]1=[13CH][13CH]=[13C]([13CH]=O)O1
Specifikationer
Kemiskt namn eller material 5-Hydroxymethyl-2-furaldehyde-13C6
Anteckningar om renhetsgrad GC
Kvantitet 10 mg
InChI formel InChI=1S/C6H6O3/c7-3-5-1-2-6(4-8)9-5/h1-3,8H,4H2/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 132.0518
Procent renhet >95%