missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxymethyl-2-furaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7020496
Click to view available options
Quantity:
1 g
5 g
25 g
67-47-0
C6H6O3
126.111
MFCD00003234
NOEGNKMFWQHSLB-UHFFFAOYSA-N
5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
237332
CHEBI:412516
5-(hydroxymethyl)furan-2-carbaldehyde
C1=C(OC(=C1)C=O)CO
67-47-0
C6H6O3
126.111
MFCD00003234
NOEGNKMFWQHSLB-UHFFFAOYSA-N
5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
237332
CHEBI:412516
5-(hydroxymethyl)furan-2-carbaldehyde
C1=C(OC(=C1)C=O)CO

5-Hydroxymethyl-2-furaldehyde is used as an intermediate of OMBF (5,5'-oxydimethylenebis(2- furfural)), crown ethers. It is used in organic synthesis to prepare dialdehydes, glycols, ethers, amino alcohols and acetals. It acts as an antioxidant found in some fruit juices and caramel products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxymethyl-2-furaldehyde is used as an intermediate of OMBF (5,5′-oxydimethylenebis(2- furfural)), crown ethers. It is used in organic synthesis to prepare dialdehydes, glycols, ethers, amino alcohols and acetals. It acts as an antioxidant found in some fruit juices and caramel products.

Solubility
Soluble in water, alcohol, ethyl acetate, acetone, dimethylformamide, benzene, ether and chloroform.

Notes
Air and light sensitive. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY
Melting Point 32°C to 35°C
Density 1.206
Boiling Point 114°C to 116°C (1 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.562
Quantity 1 g
Beilstein 110889
Sensitivity Air and light sensitive
Merck Index 14,4832
Solubility Information Soluble in water,alcohol,ethyl acetate,acetone,dimethylformamide,benzene,ether and chloroform.
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 5-Hydroxymethyl-2-furaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'