Läs mer
5-Hydroxymethyl-2-furancarboxylic Acid, TRC
Produktkod. 30391690
Change view
Kvantitet:
25 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30391690 1 g
30408713 25 mg
30408571 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30391690 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9470201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6338-41-6
Molekylformel C6 H6 O4
Molekylvikt (g/mol) 142.11
IUPAC-namn 5-(hydroxymethyl)furan-2-carboxylic acid
LEDER OCc1oc(cc1)C(=O)O
Specifikationer
Kemiskt namn eller material 5-Hydroxymethyl-2-furancarboxylic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C6H6O4/c7-3-4-1-2-5(10-4)6(8)9/h1-2,7H,3H2,(H,8,9)
Formel vikt 142.0266
Procent renhet >95%