missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Iodo-2'-Deoxycytidine, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
Specifications
Quantity | 1 g |
Formula Weight | 353.1 |
Physical Form | Solid |
Chemical Name or Material | 5-Iodo-2'-Deoxycytidine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'