Läs mer
5-Methoxy-1-naphthalenol, TRC
Produktkod. 30468617
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30468617 500 mg
30470178 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30468617 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM264705500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3588-80-5
Molekylformel C11 H10 O2
Molekylvikt (g/mol) 174.2
Synonym 1,5-Naphthohydroquinone Monomethyl Ether, 1-Hydroxy-5-methoxynaphthalene
IUPAC-namn 5-methoxynaphthalen-1-ol
LEDER COc1cccc2c(O)cccc12
Specifikationer
Kemiskt namn eller material 5-Methoxy-1-naphthalenol
Kvantitet 500 mg
InChI formel InChI=1S/C11H10O2/c1-13-11-7-3-4-8-9(11)5-2-6-10(8)12/h2-7,12H,1H3
Formel vikt 174.0681