Läs mer
5-Methoxy-2-[[(3,5-dimethylpyridin-2-yl)methyl]sulphanyl]-1H-benzimidazole, Mikromol™
Produktkod. 30051861
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051861 25 mg
30049440 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051861 Leverantör Mikromol Leverantörsnummer MM0095.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Methoxy-2-[[(3,5-dimethylpyridin-2-yl)methyl]sulphanyl]-1H-benzimidazole
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Methoxy-2-[((3,5-dimethylpyridin-2-yl)methyl)sulphanyl]-1H-benzimidazole
CAS 704910-89-4
Analyt- eller komponentnamn 5-Methoxy-2-[((3,5-dimethylpyridin-2-yl)methyl)sulphanyl]-1H-benzimidazole
Frakt skick Room Temperature
Molekylformel C16 H17 N3 O S
Certifikat och efterlevnad ISO 17025
Synonym 5-Methoxy-2-[[(3,5-dimethylpyridin-2-yl)methyl]sulphanyl]-1H-benzimidazole
InChI formel InChI=1S/C16H17N3OS/c1-10-6-11(2)15(17-8-10)9-21-16-18-13-5-4-12(20-3)7-14(13)19-16/h4-8H,9H2,1-3H3,(H,18,19)
LEDER COc1ccc2[nH]c(SCc3ncc(C)cc3C)nc2c1
IUPAC-namn 2-[(3,5-dimethylpyridin-2-yl)methylsulfanyl]-5-methoxy-1H-benzimidazole
Molekylvikt (g/mol) 299.39
Kvantitet 25 mg
Formel vikt 299.1092 g/mol