missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxypsoralen, 98%, ACROS Organics™

Brand:  Acros Organics 460660050

Additional Details : CAS Number : 484-20-8 Weight : 0.00500kg


Quantity 5g

 View more versions of this product

Product Code. 15569533

  • 1616.00 SEK / 5g


In stock, ready to ship: 1
Add to basket

Description

Description

Specifications

Chemical Identifiers

484-20-8
216.19
bergapten, 5-methoxypsoralen, bergaptene, heraclin, majudin, 4-methoxy-7h-furo 3,2-g chromen-7-one, bergaptan, psoraderm, 5-mop, o-methylbergaptol
CHEBI:18293
COC1=C2C=CC(=O)OC2=CC3=C1C=CO3
C12H8O4
BGEBZHIAGXMEMV-UHFFFAOYSA-N
2355
4-methoxyfuro[3,2-g]chromen-7-one

Specifications

5-Methoxypsoralen
2% max.
5g
97.5% min. (HPLC)
bergapten, 5-methoxypsoralen, bergaptene, heraclin, majudin, 4-methoxy-7h-furo 3,2-g chromen-7-one, bergaptan, psoraderm, 5-mop, o-methylbergaptol
COC1=C2C=CC(=O)OC2=CC3=C1C=CO3
216.19
CHEBI:18293
98%
Authentic
190.0°C to 193.0°C
484-20-8
C12H8O4
BGEBZHIAGXMEMV-UHFFFAOYSA-N
4-methoxyfuro[3,2-g]chromen-7-one
2355
216.19
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Methoxypsoralen

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Skin sensitiser Category 1

Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS
Documents