missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxypsoralen, 98%, Thermo Scientific Chemicals

Catalog Number p-7077850
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
484-20-8
C12H8O4
216.19
BGEBZHIAGXMEMV-UHFFFAOYSA-N
bergapten, 5-methoxypsoralen, bergaptene, heraclin, majudin, 4-methoxy-7h-furo 3,2-g chromen-7-one, bergaptan, psoraderm, 5-mop, o-methylbergaptol
2355
CHEBI:18293
4-methoxyfuro[3,2-g]chromen-7-one
COC1=C2C=CC(=O)OC2=CC3=C1C=CO3
This item is not returnable. View return policy
484-20-8
C12H8O4
216.19
BGEBZHIAGXMEMV-UHFFFAOYSA-N
bergapten, 5-methoxypsoralen, bergaptene, heraclin, majudin, 4-methoxy-7h-furo 3,2-g chromen-7-one, bergaptan, psoraderm, 5-mop, o-methylbergaptol
2355
CHEBI:18293
4-methoxyfuro[3,2-g]chromen-7-one
COC1=C2C=CC(=O)OC2=CC3=C1C=CO3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 190.0°C to 193.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 216.19
Percent Purity 98%
Chemical Name or Material 5-Methoxypsoralen
compliance-icons
Product Identifier
  • 5-Methoxypsoralen
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Skin sensitiser Category 1
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'