missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methoxypsoralen, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 190.0°C to 193.0°C |
Loss on Drying | 2% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 25 g |
Formula Weight | 216.19 |
Percent Purity | 98% |
Chemical Name or Material | 5-Methoxypsoralen |
Safety and Handling
Product Identifier
- 5-Methoxypsoralen
- Danger
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Skin sensitiser Category 1
- H340-May cause genetic defects.
- H350-May cause cancer.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'