Läs mer
5-Methoxyuracil, TRC
Produktkod. 30464447
Change view
Kvantitet:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464447 25 g
30471516 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30464447 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM27175025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6623-81-0
Molekylformel C5 H6 N2 O3
Molekylvikt (g/mol) 142.11
Synonym 5-Methoxy-2,4(1H,3H)-pyrimidinedione, Uracil, 5-methoxy- (6CI,7CI,8CI), 5-Methoxypyrimidine-2,4-diol, 5-Methoxyuracil, NSC 55452
IUPAC-namn 5-methoxy-1H-pyrimidine-2,4-dione
LEDER COC1=CNC(=O)NC1=O
Specifikationer
Kemiskt namn eller material 5-Methoxyuracil
Kvantitet 25 g
InChI formel InChI=1S/C5H6N2O3/c1-10-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)
Formel vikt 142.0378