Learn More
5-Methyl-1-benzothiophen-2-ylboronic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 192.05 |
Percent Purity | 97% |
Chemical Name or Material | 5-Methylbenzo[b]thiophene-2-boronic acid |