Läs mer
5-Methyl-2-[(2-nitrophenyl)amino]-3-thiophenecarbonitrile, TRC
Produktkod. 30012254
Change view
Kvantitet:
2.5 g
25 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30012254 25 g -
30008467 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012254 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32557025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 138564-59-7
Molekylformel C12 H9 N3 O2 S
Molekylvikt (g/mol) 259.28
Synonym 5-Methyl-2-[(2-nitrophenyl)amino]thiophene-3-carbonitrile, Olanzapine Imp. A (EP)
IUPAC-namn 5-methyl-2-(2-nitroanilino)thiophene-3-carbonitrile
LEDER Cc1cc(C#N)c(Nc2ccccc2[N+](=O)[O-])s1
Specifikationer
Kemiskt namn eller material 5-Methyl-2-[(2-nitrophenyl)amino]thiophene-3-carbonitrile
Kvantitet 25 g
InChI formel InChI=1S/C12H9N3O2S/c1-8-6-9(7-13)12(18-8)14-10-4-2-3-5-11(10)15(16)17/h2-6,14H,1H3
Formel vikt 259.0415