Läs mer
5-Methyl-2-hex-2-enoic Acid Ethyl Ester, TRC
Produktkod. 30468977
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30468977 1 g
1 options
Produktkod. 30468977 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3119651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Methyl-2-hex-2-enoic Acid','Ethyl Ester
CAS 34993-63-0
Molekylformel C9H16O2
Kvantitet 1 g
Synonym (E)-5-Methyl-2-Hex-2-Enoic Acid Ethyl Ester
InChI formel InChI=1S/C9H16O2/c1-4-11-9(10)7-5-6-8(2)3/h5,7-8H,4,6H2,1-3H3/b7-5+
LEDER CCOC(=O)\C=C\CC(C)C
Molekylvikt (g/mol) 156.22