Läs mer
5-Methyl-2'-O-methylcytidine, TRC
Produktkod. 30007878
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007878 100 mg
30006366 500 mg
30008534 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007878 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM263410100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 113886-70-7
Molekylformel C11 H17 N3 O5
Molekylvikt (g/mol) 271.27
Synonym 4-Amino-1-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one
IUPAC-namn 4-amino-1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxy-tetrahydrofuran-2-yl]-5-methyl-pyrimidin-2-one
LEDER CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N2C=C(C)C(=NC2=O)N
Specifikationer
Kemiskt namn eller material 5-Methyl-2'-O-methylcytidine
Kvantitet 100 mg
InChI formel InChI=1S/C11H17N3O5/c1-5-3-14(11(17)13-9(5)12)10-8(18-2)7(16)6(4-15)19-10/h3,6-8,10,15-16H,4H2,1-2H3,(H2,12,13,17)/t6-,7-,8-,10-/m1/s1
Formel vikt 271.1168