Läs mer
5-Methyl-2-phenoxypyridine, TRC
Produktkod. 30473152
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30473152 250 mg
30470546 100 mg
30478351 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30473152 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM339100250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51933-81-4
Molekylformel C12H11NO
Molekylvikt (g/mol) 185.23
IUPAC-namn 5-methyl-2-phenoxypyridine
LEDER Cc1ccc(Oc2ccccc2)nc1
Specifikationer
Kemiskt namn eller material 5-Methyl-2-phenoxypyridine
Kvantitet 250 mg
InChI formel InChI=1S/C12H11NO/c1-10-7-8-12(13-9-10)14-11-5-3-2-4-6-11/h2-9H,1H3
Formel vikt 185.0841