missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-phenyl-1,3-oxazole-4-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-3175618
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
70170-23-9
C11H9NO2
187.20
MFCD08435848
JEXONSMPSXTJFF-UHFFFAOYSA-N
5-methyl-2-phenyloxazole-4-carbaldehyde, 4-oxazolecarboxaldehyde,5-methyl-2-phenyl, 4-oxazolecarboxaldehyde, 5-methyl-2-phenyl
12471041
5-methyl-2-phenyl-1,3-oxazole-4-carbaldehyde
CC1=C(C=O)N=C(O1)C1=CC=CC=C1
This item is not returnable. View return policy
70170-23-9
C11H9NO2
187.20
MFCD08435848
JEXONSMPSXTJFF-UHFFFAOYSA-N
5-methyl-2-phenyloxazole-4-carbaldehyde, 4-oxazolecarboxaldehyde,5-methyl-2-phenyl, 4-oxazolecarboxaldehyde, 5-methyl-2-phenyl
12471041
5-methyl-2-phenyl-1,3-oxazole-4-carbaldehyde
CC1=C(C=O)N=C(O1)C1=CC=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-phenyl-1, 3-oxazole-4-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'