missing translation for 'onlineSavingsMsg'
Learn More

(5-Methyl-3-isoxazolyl)methylamine, 97+%, Thermo Scientific™

Catalog Number p-4510021
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
154016-48-5
C5H8N2O
112.132
AZVWIMLQRLKLHH-UHFFFAOYSA-N
2776306
(5-methyl-1,2-oxazol-3-yl)methanamine
CC1=CC(=NO1)CN
This item is not returnable. View return policy
154016-48-5
C5H8N2O
112.132
AZVWIMLQRLKLHH-UHFFFAOYSA-N
2776306
(5-methyl-1,2-oxazol-3-yl)methanamine
CC1=CC(=NO1)CN
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-methyl-3-isoxazolyl methylamine, 5-methylisoxazol-3-yl methanamine, 5-methyl-1,2-oxazol-3-yl methanamine, 5-methylisoxazol-3-yl methylamine, 1-5-methyl-1,2-oxazol-3-yl methanamine, 1-5-methylisoxazol-3-yl methanamine, 3-isoxazolemethanamine,5-methyl, 3-isoxazolemethanamine, 5-methyl, c-5-methyl-isoxazol-3-yl-methylamine, acmc-20a4az
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material (5-Methyl-3-isoxazolyl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'