Läs mer
5-Methyl-3-(nitromethyl)hexanoic Acid Ethyl Ester, TRC
Produktkod. 30471159
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30471159 50 mg
1 options
Produktkod. 30471159 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32444050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Methyl-3-(nitromethyl)hexanoic Acid Ethyl Ester
CAS 128013-65-0
Molekylformel C10H19NO4
Kvantitet 50 mg
Synonym Hexanoic acid','5-methyl-3-(nitromethyl)-','ethyl ester, 5-Methyl-3-nitromethylhexanoic acid ethyl ester, Ethyl 3-(nitromethyl)-5-methylhexanoate, Ethyl 5-methyl-3-(nitromethyl)hexanoate
InChI formel InChI=1S/C10H19NO4/c1-4-15-10(12)6-9(5-8(2)3)7-11(13)14/h8-9H,4-7H2,1-3H3
LEDER CCOC(=O)CC(CC(C)C)C[N+](=O)[O-]
IUPAC-namn ethyl 5-methyl-3-(nitromethyl)hexanoate
Molekylvikt (g/mol) 217.26
Formel vikt 217.1314