Learn More
5-Methyl-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carbonitrile, 97%, BLD Pharm™
Product Code. 30948434
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948434 250 mg 250mg
30948310 100 mg 100mg
30935322 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30948434 Supplier BLD Pharm Supplier No. BD00797635250mg
 Restricted Item
Beräknad leverans: 30-11-2026
Specifications
CAS 2304634-94-2
Molecular Formula C13H17BN2O2
MDL Number MFCD18715578
Quantity 250 mg
SMILES N#CC1=NC=C(C)C=C1B2OC(C)(C)C(C)(C)O2
Molecular Weight (g/mol) 244.10
Percent Purity 97%
Chemical Name or Material 5-Methyl-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carbonitrile