missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-6-(4-morpholinyl)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number 15447665
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1073372-03-8
C16H25BN2O3
304.197
MFCD09037483
DGVAEMVUOIETDY-UHFFFAOYSA-N
4-3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl morpholine, 5-methyl-6-4-morpholinyl pyridine-3-boronic acid pinacol ester, 5-methyl-6-morpholin-4-yl pyridine-3-boronic acid pinacol ester, 4-3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridyl morpholine, 5-methyl-6-morpholinylpyridine-3-boronic acid pinacol ester, 3-methyl-2-morpholinopyridine-5-boronic acid pinacol ester, 5-methyl-6-morpholinopyridine-3-boronic acid pinacol ester
44755180
4-[3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N3CCOCC3)C
This item is not returnable. View return policy
1073372-03-8
C16H25BN2O3
304.197
MFCD09037483
DGVAEMVUOIETDY-UHFFFAOYSA-N
4-3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl morpholine, 5-methyl-6-4-morpholinyl pyridine-3-boronic acid pinacol ester, 5-methyl-6-morpholin-4-yl pyridine-3-boronic acid pinacol ester, 4-3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridyl morpholine, 5-methyl-6-morpholinylpyridine-3-boronic acid pinacol ester, 3-methyl-2-morpholinopyridine-5-boronic acid pinacol ester, 5-methyl-6-morpholinopyridine-3-boronic acid pinacol ester
44755180
4-[3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N3CCOCC3)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∼100°C to 105°C
Quantity 250 mg
Formula Weight 304.2
Percent Purity 95%
Chemical Name or Material 5-Methyl-6-(4-morpholinyl)pyridine-3-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'