Läs mer
5-Methyl Cytosine-13C,15N2 Hydrochloride, TRC
Produktkod. 30009172
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009172 10 mg
30007100 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009172 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM29470210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246815-59-7
Molekylformel 13C C4 H7 15N2 N O . Cl H
Molekylvikt (g/mol) 164.57
Synonym 6-Amino-5-(methyl-13C)-2(1H)-pyrimidinone-1,3-15N2 hydrochloride, 4-Amino-2-hydroxy-5-(methyl-13C)pyrimidine-1,3-15N2 hydrochloride, 4-Amino-5-(methyl-13C)-2(1H)-pyrimidinone-1,3-15N2 hydrochloride, 5-Methyl cytosine-13C,15N2 hydrochloride, 5-Methylcytosine-13C,15N2 hydrochloride, 5-Methylcytosine-13C,15N2 (methyl-13C,1,3-15N2) hydrochloride
IUPAC-namn 4-amino-5-(1^{13}C)methyl-(1,3-^{15}N_{2})1H-pyrimidin-2-one;hydrochloride
LEDER Cl.[13CH3]C1=C[15NH]C(=O)[15N]=C1N
Specifikationer
Kemiskt namn eller material 5-Methylcytosine methyl-13C,1,3-15N2 hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C5H7N3O.ClH/c1-3-2-7-5(9)8-4(3)6;/h2H,1H3,(H3,6,7,8,9);1H/i1+1,7+1,8+1;
Formel vikt 164.033