missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-Methyl-DL-tryptophan, 98%

Catalog Number 10745221
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
951-55-3
C12H14N2O2
218.26
MFCD00005652
HUNCSWANZMJLPM-UHFFFAOYSA-N
5-methyl-dl-tryptophan, 5-methyltryptophan, 2-amino-3-5-methyl-1h-indol-3-yl propanoic acid, tryptophan, 5-methyl, dl-5-methyltryptophan, 5-methyl-tryptophan, 2-amino-3-5-methylindol-3-yl propanoic acid, 5-methyl tryptophan, dl, 5-methyltryptophan #, pubchem20652
92852
CHEBI:52524
2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid
CC1=CC=C2NC=C(CC(N)C(O)=O)C2=C1
This item is not returnable. View return policy
951-55-3
C12H14N2O2
218.26
MFCD00005652
HUNCSWANZMJLPM-UHFFFAOYSA-N
5-methyl-dl-tryptophan, 5-methyltryptophan, 2-amino-3-5-methyl-1h-indol-3-yl propanoic acid, tryptophan, 5-methyl, dl-5-methyltryptophan, 5-methyl-tryptophan, 2-amino-3-5-methylindol-3-yl propanoic acid, 5-methyl tryptophan, dl, 5-methyltryptophan #, pubchem20652
92852
CHEBI:52524
2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid
CC1=CC=C2NC=C(CC(N)C(O)=O)C2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 280.0°C to 282.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Beilstein 22,II,471
Formula Weight 218.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Methyl-DL-tryptophan
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'