Läs mer
5-Methylbenz[a]anthracene, TRC
Produktkod. 30132713
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132713 50 mg
30133155 100 mg
30007614 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132713 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM28879550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2319-96-2
Molekylformel C19H14
Molekylvikt (g/mol) 242.31
Synonym 5-Monomethylbenz[a]anthracene, 5-Methylbenzo[a]anthracene;
LEDER Cc1cc2cc3ccccc3cc2c4ccccc14
Specifikationer
Kemiskt namn eller material 5-Methylbenz[a]anthracene
Kvantitet 50 mg
InChI formel InChI=1S/C19H14/c1-13-10-16-11-14-6-2-3-7-15(14)12-19(16)18-9-5-4-8-17(13)18/h2-12H,1H3
Formel vikt 242.1096