Learn More
5-Methylcytidine, TRC
Product Code. 30482454
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482454 100 mg 100mg
30465921 5 g 5g
30479637 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30482454 Supplier Toronto Research Chemicals Supplier No. TRCM294850100MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2140-61-6
Molecular Formula C10 H15 N3 O5
Molecular Weight (g/mol) 257.24
Synonym 4-Amino-5-methyl-1-β-D-ribofuranosylpyrimidin-2(1H)-one, 5-Methylcytidine, Cytarabine Imp. G (EP)
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one
SMILES CC1=CN([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N=C1N
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C10H15N3O5/c1-4-2-13(10(17)12-8(4)11)9-7(16)6(15)5(3-14)18-9/h2,5-7,9,14-16H,3H2,1H3,(H2,11,12,17)/t5-,6-,7-,9-/m1/s1
Formula Weight 257.1012
Chemical Name or Material 5-Methylcytidine