Läs mer
5-Methylfuran-3(2H)-one, TRC
Produktkod. 30475848
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475848 100 mg
30465792 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30475848 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM236540100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Methylfuran-3(2H)-one
CAS 3511-32-8
Molekylformel C5 H6 O2
Kvantitet 100 mg
Synonym 5-Methylfuran-3(2H)-one
InChI formel InChI=1S/C5H6O2/c1-4-2-5(6)3-7-4/h2H,3H2,1H3
LEDER CC1=CC(=O)CO1
IUPAC-namn 5-methylfuran-3-one
Molekylvikt (g/mol) 98.1
Formel vikt 98.037