Läs mer
5-Methylpyrrolidin-3-ol, TRC
Produktkod. 30484151
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30484151 50 mg
30479861 10 mg
30475505 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30484151 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22248850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 94134-94-8
Molekylformel C5 H11 N O
Molekylvikt (g/mol) 101.15
IUPAC-namn 5-methylpyrrolidin-3-ol
LEDER CC1CC(O)CN1
Specifikationer
Kemiskt namn eller material 5-Methylpyrrolidin-3-ol
Kvantitet 50 mg
InChI formel InChI=1S/C5H11NO/c1-4-2-5(7)3-6-4/h4-7H,2-3H2,1H3
Formel vikt 101.0841